Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Pyridyl)pyridinium Chloride Hydrochloride (Tech.), ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5421-92-1
C10H10Cl2N2
229.104
MFCD00012821
QZOFFMRDIRXGKJ-UHFFFAOYSA-M
1-4-pyridyl pyridinium chloride hydrochloride,1,4'-bipyridin-1-ium chloride hydrochloride,n-4-pyridyl pyridinium chloride hydrochloride,1-4-pyridinyl pyridinium chloride hydrochloride,1,4'-bipyridinium, chloride,1,4'-bipyridinium, chloride, hydrochloride 1:1:1,1,4'-bipyridinium, chloride, hydrochloride,1-4-pyridyl pyridinium chloride hcl,4-pyridin-1-ium-1-ylpyridine chloride hydrochloride,4-pyridylpyridine, chloride, chloride
79461
4-pyridin-1-ium-1-ylpyridine;chloride;hydrochloride
C1=CC=[N+](C=C1)C2=CC=NC=C2.Cl.[Cl-]
5421-92-1
C10H10Cl2N2
229.104
MFCD00012821
QZOFFMRDIRXGKJ-UHFFFAOYSA-M
1-4-pyridyl pyridinium chloride hydrochloride,1,4'-bipyridin-1-ium chloride hydrochloride,n-4-pyridyl pyridinium chloride hydrochloride,1-4-pyridinyl pyridinium chloride hydrochloride,1,4'-bipyridinium, chloride,1,4'-bipyridinium, chloride, hydrochloride 1:1:1,1,4'-bipyridinium, chloride, hydrochloride,1-4-pyridyl pyridinium chloride hcl,4-pyridin-1-ium-1-ylpyridine chloride hydrochloride,4-pyridylpyridine, chloride, chloride
79461
4-pyridin-1-ium-1-ylpyridine;chloride;hydrochloride
C1=CC=[N+](C=C1)C2=CC=NC=C2.Cl.[Cl-]
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Brown
Formula Weight 229.11
Physical Form Powder
Percent Purity ≥70% (typically)
Grade Technical
Chemical Name or Material N-(4-Pyridyl)pyridinium chloride hydrochloride, tech.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 226-543-5

TSCA : TSCA