Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(5-Methoxy-1H-indol-3-yl)methyl]-N,N-dimethylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
16620-52-3
C12H16N2O
204.273
GOERTRUXQHDLHC-UHFFFAOYSA-N
5-methoxygramine,methoxygramine,1-5-methoxy-1h-indol-3-yl-n,n-dimethylmethanamine,1h-indole-3-methanamine, 5-methoxy-n,n-dimethyl,3-dimethylaminomethyl-5-methoxyindole,2-dimethylaminomethyl-5-methoxyindole,5-methoxy-3-dimethylaminomethyl indole,indole, 3-dimethylamino methyl-5-methoxy,n-5-methoxy-1h-indol-3-yl methyl-n,n-dimethylamine
27965
1-(5-methoxy-1H-indol-3-yl)-N,N-dimethylmethanamine
CN(C)CC1=CNC2=C1C=C(C=C2)OC
16620-52-3
C12H16N2O
204.273
GOERTRUXQHDLHC-UHFFFAOYSA-N
5-methoxygramine,methoxygramine,1-5-methoxy-1h-indol-3-yl-n,n-dimethylmethanamine,1h-indole-3-methanamine, 5-methoxy-n,n-dimethyl,3-dimethylaminomethyl-5-methoxyindole,2-dimethylaminomethyl-5-methoxyindole,5-methoxy-3-dimethylaminomethyl indole,indole, 3-dimethylamino methyl-5-methoxy,n-5-methoxy-1h-indol-3-yl methyl-n,n-dimethylamine
27965
1-(5-methoxy-1H-indol-3-yl)-N,N-dimethylmethanamine
CN(C)CC1=CNC2=C1C=C(C=C2)OC
Infrared Spectrum Authentic
Melting Point 125°C to 127°C
Quantity 1g
Packaging Glass bottle
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 204.27
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Methoxygramine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger