Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(6-Bromohexyl)phthalimide, 97%

Click to view available options
Quantity:
5g
25g
100g
24566-79-8
C14H16BrNO2
310.191
MFCD00023098
OAZFTIPKNPTDIO-UHFFFAOYSA-N
n-6-bromohexyl phthalimide,2-6-bromohexyl isoindoline-1,3-dione,n-6-bromohexyl-phthalimide,2-6-bromohexyl-1h-isoindole-1,3 2h-dione,2-6-bromohexyl isoindole-1,3-dione,2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione,2-6-bromo-hexyl-isoindole-1,3-dione,1h-isoindole-1,3 2h-dione,2-6-bromohexyl,1h-isoindole-1,3 2h-dione, 2-6-bromohexyl,6-n-phthalimido hexyl bromide
141120
2-(6-bromohexyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr
24566-79-8
C14H16BrNO2
310.191
MFCD00023098
OAZFTIPKNPTDIO-UHFFFAOYSA-N
n-6-bromohexyl phthalimide,2-6-bromohexyl isoindoline-1,3-dione,n-6-bromohexyl-phthalimide,2-6-bromohexyl-1h-isoindole-1,3 2h-dione,2-6-bromohexyl isoindole-1,3-dione,2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione,2-6-bromo-hexyl-isoindole-1,3-dione,1h-isoindole-1,3 2h-dione,2-6-bromohexyl,1h-isoindole-1,3 2h-dione, 2-6-bromohexyl,6-n-phthalimido hexyl bromide
141120
2-(6-bromohexyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr
Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
Odor Odorless
Boiling Point 186°C to 188°C (0.4mmHg)
Melting Point 54°C to 60°C
Quantity 5g
Beilstein 186875
Formula Weight 310.19
Percent Purity 97%
Chemical Name or Material N-(6-Bromohexyl)phthalimide