Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, TCI America™

Click to view available options
Quantity:
1g
5g
135112-28-6
C20H21NO4
339.391
MFCD00155631
JBIJSEUVWWLFGV-SFHVURJKSA-N
fmoc-nva-oh,fmoc-l-norvaline,n-9-fluorenylmethyloxycarbonyl-l-norvaline,n-fmoc-l-norvaline,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid,l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl,fmoc-norvaline,fmoc-l-nva-oh,ambotzfaa1415,pubchem14961
7016885
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
135112-28-6
C20H21NO4
339.391
MFCD00155631
JBIJSEUVWWLFGV-SFHVURJKSA-N
fmoc-nva-oh,fmoc-l-norvaline,n-9-fluorenylmethyloxycarbonyl-l-norvaline,n-fmoc-l-norvaline,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid,l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl,fmoc-norvaline,fmoc-l-nva-oh,ambotzfaa1415,pubchem14961
7016885
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Color White
Melting Point 155°C
Quantity 5g
Formula Weight 339.39
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline

TSCA : No

Recommended Storage : Refrigerator