Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline, TCI America™

Click to view available options
Quantity:
1g
5g
252049-05-1
C21H23NO4
353.418
MFCD01861326
HKELUUGCKFRJQM-IBGZPJMESA-N
fmoc-n-methyl-l-norvaline,fmoc-n-me-nva-oh,s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino pentanoic acid,fmoc-n-me-l-2-aminovaleric acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino pentanoic acid,l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl,l-norvaline,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl,fmoc-n-me-l-norvaline,n-fmoc-n-methyl-l-norvaline,fmoc-nalpha-methyl-l-norvaline
7009909
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]pentanoic acid
CCCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
252049-05-1
C21H23NO4
353.418
MFCD01861326
HKELUUGCKFRJQM-IBGZPJMESA-N
fmoc-n-methyl-l-norvaline,fmoc-n-me-nva-oh,s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino pentanoic acid,fmoc-n-me-l-2-aminovaleric acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino pentanoic acid,l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl,l-norvaline,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl,fmoc-n-me-l-norvaline,n-fmoc-n-methyl-l-norvaline,fmoc-nalpha-methyl-l-norvaline
7009909
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]pentanoic acid
CCCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Melting Point 132°C
Quantity 5g
Color White
Formula Weight 353.42
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline