Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methylglycine, TCI America™

Click to view available options
Quantity:
5g
25g
77128-70-2
C18H19NO5
329.352
MFCD00151926
CUJSWOOWOONPRH-UHFFFAOYSA-N
fmoc-sarcosine monohydrate,fmoc-sarcosine hydrate,2-9h-fluoren-9-yl methoxy carbonyl methyl amino acetic acid hydrate,9h-fluoren-9-ylmethoxy carbonyl methyl amino acetic acid hydrate,2-9h-fluoren-9-ylmethoxycarbonyl methyl amino acetic acid;hydrate,fmoc-sarcosinehydrate,ksc915q4d,n-9h-fluoren-9-yl methoxy carbonyl-n-methylglycine-water 1/1
56763849
2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]acetic acid;hydrate
CN(CC(=O)O)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O
77128-70-2
C18H19NO5
329.352
MFCD00151926
CUJSWOOWOONPRH-UHFFFAOYSA-N
fmoc-sarcosine monohydrate,fmoc-sarcosine hydrate,2-9h-fluoren-9-yl methoxy carbonyl methyl amino acetic acid hydrate,9h-fluoren-9-ylmethoxy carbonyl methyl amino acetic acid hydrate,2-9h-fluoren-9-ylmethoxycarbonyl methyl amino acetic acid;hydrate,fmoc-sarcosinehydrate,ksc915q4d,n-9h-fluoren-9-yl methoxy carbonyl-n-methylglycine-water 1/1
56763849
2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]acetic acid;hydrate
CN(CC(=O)O)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O
Quantity 25g
Melting Point 119°C
Color White
Formula Weight 311.34
Physical Form Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methylglycine