Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-1-aminonaphthalene, 98%, Acros Organics

Click to view available options
Quantity:
5g
575-36-0
C12H11NO
185.226
MFCD00059128
OKQIEBVRUGLWOR-UHFFFAOYSA-N
1-acetamidonaphthalene,acetamide, n-1-naphthalenyl,n-1-naphthylacetamide,n-naphthalen-1-yl acetamide,n-1-naphthyl acetamide,n-acetyl-1-naphthylamine,n-acetyl-1-aminonaphthalene,n-naphthylacetamide,acetamide, n-1-naphthyl
68461
N-naphthalen-1-ylacetamide
CC(=O)NC1=CC=CC2=CC=CC=C21
575-36-0
C12H11NO
185.226
MFCD00059128
OKQIEBVRUGLWOR-UHFFFAOYSA-N
1-acetamidonaphthalene,acetamide, n-1-naphthalenyl,n-1-naphthylacetamide,n-naphthalen-1-yl acetamide,n-1-naphthyl acetamide,n-acetyl-1-naphthylamine,n-acetyl-1-aminonaphthalene,n-naphthylacetamide,acetamide, n-1-naphthyl
68461
N-naphthalen-1-ylacetamide
CC(=O)NC1=CC=CC2=CC=CC=C21
Infrared Spectrum Authentic
Color Violet
Quantity 5g
Melting Point 156°C to 160°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 185.22
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material N-Acetyl-1-aminonaphthalene, 98%
Show More Show Less

EINECSNumber : 209-380-4

TSCA : TSCA