Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-(2R)-bornane-10,2-sultam, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
141993-16-0
C12H19NO3S
257.35
MFCD00269649
FECARJHVTMSYSJ-FOGDFJRCSA-N
n-acetyl-2r-bornane-10,2-sultam
60161326
CC(=O)N1C2CC3CCC2(C3(C)C)CS1(=O)=O
141993-16-0
C12H19NO3S
257.35
MFCD00269649
FECARJHVTMSYSJ-FOGDFJRCSA-N
n-acetyl-2r-bornane-10,2-sultam
60161326
CC(=O)N1C2CC3CCC2(C3(C)C)CS1(=O)=O
Quantity 1g
Packaging Glass bottle
Formula Weight 257.35
Chemical Name or Material N-Acetyl-(2R)-bornane-10, 2-sultam

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning