Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-3-(2-naphthyl)-D-alanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
37440-01-0
C15H15NO3
257.289
MFCD00079678
HGTIILKZSFKZMS-CQSZACIVSA-N
ac-d-2-nal-oh,r-2-acetamido-3-naphthalen-2-yl propanoic acid,r-n-acetyl-2-naphthylalanine,2r-2-acetamido-3-naphthalen-2-yl propanoic acid,pubchem12668,n-acetyl-3-2-naphthyl-d-alanine,2r-2-acetamido-3-naphthalen-2-ylpropanoic acid,r-2-acetamido-3-naphthalen-2-yl propanoicacid,2r-2-acetylamino-3-2-naphthyl propanoic acid,2s-2-acetylamino-3-naphth-2-yl propanoic acid
1268231
(2R)-2-acetamido-3-naphthalen-2-ylpropanoic acid
CC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O
37440-01-0
C15H15NO3
257.289
MFCD00079678
HGTIILKZSFKZMS-CQSZACIVSA-N
ac-d-2-nal-oh,r-2-acetamido-3-naphthalen-2-yl propanoic acid,r-n-acetyl-2-naphthylalanine,2r-2-acetamido-3-naphthalen-2-yl propanoic acid,pubchem12668,n-acetyl-3-2-naphthyl-d-alanine,2r-2-acetamido-3-naphthalen-2-ylpropanoic acid,r-2-acetamido-3-naphthalen-2-yl propanoicacid,2r-2-acetylamino-3-2-naphthyl propanoic acid,2s-2-acetylamino-3-naphth-2-yl propanoic acid
1268231
(2R)-2-acetamido-3-naphthalen-2-ylpropanoic acid
CC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O
Beilstein 7193723
Formula Weight 257.29
Physical Form Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material N-Acetyl-3-(2-naphthyl)-D-alanine

storageNote1 : -30° to -10°C