Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-Adhesin (1025-1044) amide

Click to view available options
Quantity:
0.5mg
1mg
MFCD02262418
N-Acetyl-Gln-Leu-Lys-Thr-Ala-Asp-Leu-Pro-Ala-Gly-Arg-Asp-Glu-Thr-Thr-Ser-Phe-Val-Leu-Val-NH2; p1025
MFCD02262418
N-Acetyl-Gln-Leu-Lys-Thr-Ala-Asp-Leu-Pro-Ala-Gly-Arg-Asp-Glu-Thr-Thr-Ser-Phe-Val-Leu-Val-NH2; p1025
Quantity 1mg
Color White
Formula Weight 2202.46
Physical Form Lyophilized Powder
Chemical Name or Material N-Acetyl-Adhesin (1025-1044) amide

storageNote1 : -30° to -10°C