Learn More
Alfa Aesar™ N-Acetyl-Asp-Glu-Val-Asp-al
Manufacturer:  Alfa Aesar™ J65204MCR
 View more versions of this product
Catalog No. AAJ65204MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1mg
-30°C to -10°C
N-Acetyl-Asp-Glu-Val-Asp-al
169332-60-9
502.47
Moisture Sensitive
White
Powder
Reversible inhibitor of interleukin-1beta converting enzyme (ICE); also inhibits caspase-3 cleavage of poly(ADP-ribose) polymerase
C20H30N4O11
Ac-Asp-Glu-Val-Asp-CHO; Ac-DEVD-CHO