Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl Cholecystokinin, CCK (26-30), sulfated

Click to view available options
Quantity:
0.5mg
1mg
MFCD00133563
N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-NH2
MFCD00133563
N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-NH2
Quantity 0.5mg
Color White
Formula Weight 791.85
Physical Form Solid
Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-30), sulfated

storageNote1 : -30° to -10°C