Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-galactosamine Hydrate 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
14215-68-0
C8H15NO6
221.209
MFCD00065372
OVRNDRQMDRJTHS-JAJWTYFOSA-N
n-acetyl-d-galactosamine,galnac,2-acetamido-2-deoxy-d-galactopyranose,d-galnac,2-acetamido-2-deoxy-d-galactose,n-acetyl-d-chondrosamine,n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide,n-acetylgalactosamine,acetylgalactosamine,n-acetyl-galactosamine
440552
CHEBI:28497
N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
CC(=O)NC1C(C(C(OC1O)CO)O)O
14215-68-0
C8H15NO6
221.209
MFCD00065372
OVRNDRQMDRJTHS-JAJWTYFOSA-N
n-acetyl-d-galactosamine,galnac,2-acetamido-2-deoxy-d-galactopyranose,d-galnac,2-acetamido-2-deoxy-d-galactose,n-acetyl-d-chondrosamine,n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide,n-acetylgalactosamine,acetylgalactosamine,n-acetyl-galactosamine
440552
CHEBI:28497
N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
CC(=O)NC1C(C(C(OC1O)CO)O)O
Color White-Yellow
Quantity 1g
Formula Weight 221.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material N-Acetyl-D-galactosamine Hydrate