Learn More
Diazyme Laboratories N-Acetyl-β-D-Glucosaminidase (NAG) Assay
N-Acetyl-β-D-glucosaminidase (NAG) Assay Kit is for determination of NAG in urine samples.
Manufacturer:  Diazyme Laboratories DZ062AK
Catalog No. 23-700-574
Add to cart
Description
Description
  • Ready-to-use for manual methods and adaptable for analyzers
  • The assay is specific for NAG and has excellent sensitivity with a linear range up to 200 IU/L² and excellent precision with a CV% of <5%
  • There is no significant interference from hemoglobin or albumin
Specifications
Specifications
USA: For Research Use Only, EU: CE IVD
R1: Stable for 1 week when stored capped at 2 to 8°C; R3: Stable until the expiration dated indicated on its label when unopened and stored at 2 to 8°C
Yes
N-Acetyl-β-D-Glucosaminidase (NAG) Assay Kit
Eleven levels of linearity set were prepared by diluting a sample containing 200 U/L NAG with a low sample containing 10.93 U/L NAG according to Clinical and Laboratory Standards Institute (for- merly NCCLS) EP6-A and tested on Hitachi 917. The Diazyme NAG Assay is linear up to 200 U/L.
R1: Stable for 1 week when stored capped at 2 to 8°C; R3: Stable until the expiration dated indicated on its label when unopened and stored at 2 to 8°C
R1: 1 x 75 mL; R2: 1 x 15 mL; R3: 1 x 30 mL; Cal: 1 x 2 mL vial, lyophilized (Calibrator Included and Packaged Separately)
Diagnostic
Lab dependent
Reagents
Universal