Alfa Aesar™ N-Acetyl-D-leucine, 99%

Manufacturer: Alfa Aesar™ H2704603

Quantity 1g

 View more versions of this product

Catalog No. AAH2704603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19764-30-8
    Molecular Formula C8H15NO3
    Formula Weight 173.21
    MDL Number MFCD00066069
    Synonym Ac-D-Leu-OH
    Chemical Name or Material N-Acetyl-D-leucine

    Specifications

    Formula Weight 173.21
    Molecular Formula C8H15NO3
    MDL Number MFCD00066069
    CAS 19764-30-8
    Synonym Ac-D-Leu-OH
    Quantity 1g
    Recommended Storage −30° to −10°C
    Chemical Name or Material N-Acetyl-D-leucine
    Assay 99%

    −30° to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit