Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-methionine, 98%, Thermo Scientific™

Catalog Number AAH6680822
Click to view available options
Quantity:
5 g
25 g
100 g
1509-92-8
C7H13NO3S
191.25
MFCD00069962
XUYPXLNMDZIRQH-ZCFIWIBFSA-N
n-acetyl-d-methionine, ac-d-met-oh, n-acetylmethionine, d, n-acetyl-d-met, unii-14d624n0ta, acetyl-d-methionine, ac-d-methionine, n-ac-d-methionine, zlchem 335
6991987
CHEBI:85210
(2R)-2-acetamido-4-(methylsulfanyl)butanoic acid
CSCC[C@@H](NC(C)=O)C(O)=O
1509-92-8
C7H13NO3S
191.25
MFCD00069962
XUYPXLNMDZIRQH-ZCFIWIBFSA-N
n-acetyl-d-methionine, ac-d-met-oh, n-acetylmethionine, d, n-acetyl-d-met, unii-14d624n0ta, acetyl-d-methionine, ac-d-methionine, n-ac-d-methionine, zlchem 335
6991987
CHEBI:85210
(2R)-2-acetamido-4-(methylsulfanyl)butanoic acid
CSCC[C@@H](NC(C)=O)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 104°C
Color White
Beilstein 1725553
Formula Weight 191.25
Percent Purity 98%
Quantity 100 g
Physical Form Crystalline
Chemical Name or Material N-Acetyl-D-methionine
EINECSNumber 216-144-4
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit