Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-D-methionine, 98%

Click to view available options
Quantity:
5g
25g
100g
1509-92-8
C7H13NO3S
191.245
MFCD00069962
XUYPXLNMDZIRQH-ZCFIWIBFSA-N
n-acetyl-d-methionine,ac-d-met-oh,n-acetylmethionine, d,n-acetyl-d-met,unii-14d624n0ta,n-acetyl-d-methionine,acetyl-d-methionine,ac-d-methionine,n-ac-d-methionine,zlchem 335
6991987
CHEBI:85210
(2R)-2-acetamido-4-methylsulfanylbutanoic acid
CC(=O)NC(CCSC)C(=O)O
1509-92-8
C7H13NO3S
191.245
MFCD00069962
XUYPXLNMDZIRQH-ZCFIWIBFSA-N
n-acetyl-d-methionine,ac-d-met-oh,n-acetylmethionine, d,n-acetyl-d-met,unii-14d624n0ta,n-acetyl-d-methionine,acetyl-d-methionine,ac-d-methionine,n-ac-d-methionine,zlchem 335
6991987
CHEBI:85210
(2R)-2-acetamido-4-methylsulfanylbutanoic acid
CC(=O)NC(CCSC)C(=O)O
Beilstein 1725553
Formula Weight 191.25
Physical Form Crystalline
Percent Purity 98%
Color White
Melting Point 102°C to 104°C
Quantity 5g
Chemical Name or Material N-Acetyl-D-methionine

storageNote1 : -30° to -10°C