Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-D-proline, 98%

Click to view available options
Quantity:
5g
25g
59785-68-1
C7H11NO3
157.169
MFCD00080841
GNMSLDIYJOSUSW-ZCFIWIBFSA-N
n-acetyl-d-proline,ac-d-pro-oh,d-proline, 1-acetyl,r-1-acetylpyrrolidine-2-carboxylic acid,1-acetyl-d-proline,2r-1-acetylpyrrolidine-2-carboxylic acid,4ayu,ambotzaaa1931,ac-d-prn-oh,r-+-acetylproline
719436
CHEBI:44272
(2R)-1-acetylpyrrolidine-2-carboxylic acid
CC(=O)N1CCCC1C(=O)O
59785-68-1
C7H11NO3
157.169
MFCD00080841
GNMSLDIYJOSUSW-ZCFIWIBFSA-N
n-acetyl-d-proline,ac-d-pro-oh,d-proline, 1-acetyl,r-1-acetylpyrrolidine-2-carboxylic acid,1-acetyl-d-proline,2r-1-acetylpyrrolidine-2-carboxylic acid,4ayu,ambotzaaa1931,ac-d-prn-oh,r-+-acetylproline
719436
CHEBI:44272
(2R)-1-acetylpyrrolidine-2-carboxylic acid
CC(=O)N1CCCC1C(=O)O
Formula Weight 157.17
Physical Form Solid
Percent Purity 98%
Quantity 25g
Chemical Name or Material N-Acetyl-D-proline

storageNote1 : -30° to -10°C