Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-proline, 98%, Thermo Scientific™

Catalog Number AAH6652406
Click to view available options
Quantity:
5 g
25 g
59785-68-1
C7H11NO3
157.169
MFCD00080841
GNMSLDIYJOSUSW-ZCFIWIBFSA-N
n-acetyl-d-proline, ac-d-pro-oh, d-proline, 1-acetyl, r-1-acetylpyrrolidine-2-carboxylic acid, 1-acetyl-d-proline, 2r-1-acetylpyrrolidine-2-carboxylic acid, 4ayu, ambotzaaa1931, ac-d-prn-oh, r-+-acetylproline
719436
CHEBI:44272
(2R)-1-acetylpyrrolidine-2-carboxylic acid
CC(=O)N1CCCC1C(=O)O
59785-68-1
C7H11NO3
157.169
MFCD00080841
GNMSLDIYJOSUSW-ZCFIWIBFSA-N
n-acetyl-d-proline, ac-d-pro-oh, d-proline, 1-acetyl, r-1-acetylpyrrolidine-2-carboxylic acid, 1-acetyl-d-proline, 2r-1-acetylpyrrolidine-2-carboxylic acid, 4ayu, ambotzaaa1931, ac-d-prn-oh, r-+-acetylproline
719436
CHEBI:44272
(2R)-1-acetylpyrrolidine-2-carboxylic acid
CC(=O)N1CCCC1C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 157.17
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Acetyl-D-proline
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit