Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-D-valine, 98%

Click to view available options
Quantity:
5g
25g
100g
17916-88-0
C7H13NO3
159.185
MFCD03939895
IHYJTAOFMMMOPX-ZCFIWIBFSA-N
n-acetyl-d-valine,ac-d-val-oh,d-valine, n-acetyl,acetyl-d-valine,2r-2-acetamido-3-methylbutanoic acid,n-acetylvaline,2r-2-acetylamino-3-methylbutanoic acid,n-acetylvaline #
76475
(2R)-2-acetamido-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)C
17916-88-0
C7H13NO3
159.185
MFCD03939895
IHYJTAOFMMMOPX-ZCFIWIBFSA-N
n-acetyl-d-valine,ac-d-val-oh,d-valine, n-acetyl,acetyl-d-valine,2r-2-acetamido-3-methylbutanoic acid,n-acetylvaline,2r-2-acetylamino-3-methylbutanoic acid,n-acetylvaline #
76475
(2R)-2-acetamido-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)C
Formula Weight 159.19
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material N-Acetyl-D-valine

storageNote1 : -30° to -10°C