Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-DL-histidine hydrate, 98%

Click to view available options
Quantity:
1g
5g
213178-97-3
C8H13N3O4
215.209
MFCD00150969
PSWSDQRXCOJSFC-UHFFFAOYSA-N
2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate,ac-dl-his-oh.h2o,n-acetyl-dl-histidine monohydrate,2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate,2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate,n-acetyl-l-histidine monohydrate,2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate,acmc-1c6iz,acmc-209j3m,n-acetyl-dl-histidine hydrate
18668507
2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
CC(=O)NC(CC1=CN=CN1)C(=O)O.O
213178-97-3
C8H13N3O4
215.209
MFCD00150969
PSWSDQRXCOJSFC-UHFFFAOYSA-N
2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate,ac-dl-his-oh.h2o,n-acetyl-dl-histidine monohydrate,2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate,2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate,n-acetyl-l-histidine monohydrate,2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate,acmc-1c6iz,acmc-209j3m,n-acetyl-dl-histidine hydrate
18668507
2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
CC(=O)NC(CC1=CN=CN1)C(=O)O.O
Formula Weight 197.19 (Anhydrous)
Physical Form Solid
Percent Purity 98%
Quantity 1g
Chemical Name or Material N-Acetyl-DL-histidine hydrate