Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-DL-norvaline, 98%

Click to view available options
Quantity:
1g
5g
25g
7682-15-7
C7H13NO3
159.185
MFCD00037269
BSYFPUSAWVWWDG-UHFFFAOYSA-N
n-acetyl-dl-norvaline,ac-dl-nva-oh,acetyl-dl-norvaline,n-acetylnorvaline #,2-acetamido-pentanoic acid,2-acetylamino valeric acid,n-alpha-acetyl-dl-norvaline,n-acetyl-2-aminovaleric acid,2-acetylamino pentanoic acid
306106
2-acetamidopentanoic acid
CCCC(C(=O)O)NC(=O)C
7682-15-7
C7H13NO3
159.185
MFCD00037269
BSYFPUSAWVWWDG-UHFFFAOYSA-N
n-acetyl-dl-norvaline,ac-dl-nva-oh,acetyl-dl-norvaline,n-acetylnorvaline #,2-acetamido-pentanoic acid,2-acetylamino valeric acid,n-alpha-acetyl-dl-norvaline,n-acetyl-2-aminovaleric acid,2-acetylamino pentanoic acid
306106
2-acetamidopentanoic acid
CCCC(C(=O)O)NC(=O)C
Formula Weight 159.19
Percent Purity 98%
Quantity 5g
Chemical Name or Material N-Acetyl-DL-norvaline