Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-DL-phenylglycine, 98%

Click to view available options
Quantity:
5g
25g
100g
15962-46-6
C10H11NO3
193.202
MFCD00055924
VKDFZMMOLPIWQQ-UHFFFAOYSA-N
acetylamino phenyl acetic acid,ac-dl-phg-oh,2-acetylamino-2-phenylacetic acid,acetamido phenyl acetic acid,n-acetyl-l-phenylglycine,n-acetyl-dl-2-phenylglycine,1-acetamido phenylacetic acid,phenylaceturic acid,n-acetyl-2-phenylglycine,maybridge1_002435
85935
2-acetamido-2-phenylacetic acid
CC(=O)NC(C1=CC=CC=C1)C(=O)O
15962-46-6
C10H11NO3
193.202
MFCD00055924
VKDFZMMOLPIWQQ-UHFFFAOYSA-N
acetylamino phenyl acetic acid,ac-dl-phg-oh,2-acetylamino-2-phenylacetic acid,acetamido phenyl acetic acid,n-acetyl-l-phenylglycine,n-acetyl-dl-2-phenylglycine,1-acetamido phenylacetic acid,phenylaceturic acid,n-acetyl-2-phenylglycine,maybridge1_002435
85935
2-acetamido-2-phenylacetic acid
CC(=O)NC(C1=CC=CC=C1)C(=O)O
Formula Weight 193.2
Physical Form Powder
Percent Purity 98%
Color White
Quantity 100g
Chemical Name or Material N-Acetyl-DL-phenylglycine

storageNote1 : -30° to -10°C