Alfa Aesar™ N-Acetyl-DL-serine, 98+%

Manufacturer: Alfa Aesar™ H2771714

Quantity 25g

 View more versions of this product

Catalog No. AAH2771714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97-14-3
    Molecular Formula C5H9NO4
    Formula Weight 147.13
    MDL Number MFCD00037248
    Chemical Name or Material N-Acetyl-DL-serine

    Specifications

    Formula Weight 147.13
    Molecular Formula C5H9NO4
    MDL Number MFCD00037248
    CAS 97-14-3
    Quantity 25g
    Recommended Storage −30° to −10°C
    Chemical Name or Material N-Acetyl-DL-serine
    Assay ≥98%

    −30° to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit