Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ N-Acetyl-Endothelin-1 (16-21), Human

For cellular and molecular biology applications

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 837.96
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ66512EXD
View Documents
Thermo Scientific™
J66512EXD
0.5 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
AAJ66512MCR
View Documents
Thermo Scientific™
J66512MCR
1 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Human
-30°C to -10°C
837.96
RUO - research use only
HPLC
Solid
N-Acetyl-Endothelin-1 (16-21), human
N-Acetyl-His-Leu-Asp-Ile-Ile-Trp
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit