Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-Ile-Glu-Pro-Asp-al

Click to view available options
Quantity:
1mg
5mg
MFCD02259689
Ac-IEPD-CHO; Caspase-8 Inhibitor
MFCD02259689
Ac-IEPD-CHO; Caspase-8 Inhibitor
Quantity 5mg
Color White
Formula Weight 498.53
Physical Form Lyophilized Powder
Chemical Name or Material N-Acetyl-Ile-Glu-Pro-Asp-al

storageNote1 : -30° to -10°C