Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-Ile-Glu-Thr-Asp-al

Click to view available options
Quantity:
1mg
5mg
191338-86-0
C21H34N4O10
502.521
MFCD01318858
AXTKTZHLZLOIIO-PBEPODTISA-N
ac-ietd-cho,ac-ile-glu-thr-asp-aldehyde pseudo acid,4s-4-1s,2r-1-2s-1-carboxy-3-oxopropan-2-yl carbamoyl-2-hydroxypropyl carbamoyl-4-2s,3s-2-acetamido-3-methylpentanamido butanoic acid,ac-ile-glu-thr-asp-cho,acetyl-ile-glu-thr-asp-aldehyde,d0n8ta,acetyl-isoleucyl-glutamyl-threonyl-aspartal,n-acetyl-ile-glu-thr-asp-al tlc , powder,4s,7s,10s,13s-4-sec-butyl-7-2-carboxyethyl-13-formyl-10-r-1-hydroxyethyl-2,5,8,11-tetraoxo-3,6,9,12-tetraazapentadecan-15-oic acid
11466200
(4S)-4-[[(2S,3S)-2-acetamido-3-methylpentanoyl]amino]-5-[[(2S,3R)-1-[[(2S)-1-carboxy-3-oxopropan-2-yl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-5-oxopentanoic acid
CCC(C)C(C(=O)NC(CCC(=O)O)C(=O)NC(C(C)O)C(=O)NC(CC(=O)O)C=O)NC(=O)C
191338-86-0
C21H34N4O10
502.521
MFCD01318858
AXTKTZHLZLOIIO-PBEPODTISA-N
ac-ietd-cho,ac-ile-glu-thr-asp-aldehyde pseudo acid,4s-4-1s,2r-1-2s-1-carboxy-3-oxopropan-2-yl carbamoyl-2-hydroxypropyl carbamoyl-4-2s,3s-2-acetamido-3-methylpentanamido butanoic acid,ac-ile-glu-thr-asp-cho,acetyl-ile-glu-thr-asp-aldehyde,d0n8ta,acetyl-isoleucyl-glutamyl-threonyl-aspartal,n-acetyl-ile-glu-thr-asp-al tlc , powder,4s,7s,10s,13s-4-sec-butyl-7-2-carboxyethyl-13-formyl-10-r-1-hydroxyethyl-2,5,8,11-tetraoxo-3,6,9,12-tetraazapentadecan-15-oic acid
11466200
(4S)-4-[[(2S,3S)-2-acetamido-3-methylpentanoyl]amino]-5-[[(2S,3R)-1-[[(2S)-1-carboxy-3-oxopropan-2-yl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-5-oxopentanoic acid
CCC(C)C(C(=O)NC(CCC(=O)O)C(=O)NC(C(C)O)C(=O)NC(CC(=O)O)C=O)NC(=O)C
Quantity 1mg
Color White
Formula Weight 502.51
Physical Form Lyophilized Powder
Chemical Name or Material N-Acetyl-Ile-Glu-Thr-Asp-al

storageNote1 : -30° to -10°C