Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-L-alanine methyl ester, 95%

Click to view available options
Quantity:
5g
25g
3637-01-2
C10H12O
148.21
MFCD03940666
WPRAXAOJIODQJR-UHFFFAOYSA-N
3',4'-dimethylacetophenone,1-3,4-dimethylphenyl ethanone,3,4-dimethylacetophenone,ethanone, 1-3,4-dimethylphenyl,1-3,4-dimethylphenyl ethan-1-one,unii-pc1l145t5v,1-3,4-dimethyl-phenyl-ethanone,1-acetyl-3,4-dimethylbenzene,3.4-dimethylacetophenon,acmc-1cmy2
77193
1-(3,4-dimethylphenyl)ethanone
CC1=C(C=C(C=C1)C(=O)C)C
3637-01-2
C10H12O
148.21
MFCD03940666
WPRAXAOJIODQJR-UHFFFAOYSA-N
3',4'-dimethylacetophenone,1-3,4-dimethylphenyl ethanone,3,4-dimethylacetophenone,ethanone, 1-3,4-dimethylphenyl,1-3,4-dimethylphenyl ethan-1-one,unii-pc1l145t5v,1-3,4-dimethyl-phenyl-ethanone,1-acetyl-3,4-dimethylbenzene,3.4-dimethylacetophenon,acmc-1cmy2
77193
1-(3,4-dimethylphenyl)ethanone
CC1=C(C=C(C=C1)C(=O)C)C
Beilstein 1723226
Formula Weight 145.16
Physical Form Liquid
Percent Purity 95%
Color Yellow
Quantity 25g
Chemical Name or Material N-Acetyl-L-alanine methyl ester

storageNote1 : -30° to -10°C