Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-L-alanine methyl ester, 95%

Click to view available options
Quantity:
5g
25g
3619-02-1
C6H11NO3
145.16
MFCD03940666
FQGVVDYNRHNTCK-UHFFFAOYNA-N
s-methyl 2-acetamidopropanoate,ac-ala-ome,n-acetyl-l-alanine methyl ester,methyl 2s-2-acetamidopropanoate,l-alanine, n-acetyl-, methyl ester,alanine, n-acetyl-,methyl ester,ac-l-ala-ome,ambotzaaa1912,n-acetyl-l-alanine methyl,n-acetylalanine methyl ester
6951030
methyl (2S)-2-acetamidopropanoate
CC(C(=O)OC)NC(=O)C
3619-02-1
C6H11NO3
145.16
MFCD03940666
FQGVVDYNRHNTCK-UHFFFAOYNA-N
s-methyl 2-acetamidopropanoate,ac-ala-ome,n-acetyl-l-alanine methyl ester,methyl 2s-2-acetamidopropanoate,l-alanine, n-acetyl-, methyl ester,alanine, n-acetyl-,methyl ester,ac-l-ala-ome,ambotzaaa1912,n-acetyl-l-alanine methyl,n-acetylalanine methyl ester
6951030
methyl (2S)-2-acetamidopropanoate
CC(C(=O)OC)NC(=O)C
Beilstein 1723226
Formula Weight 145.16
Physical Form Liquid
Percent Purity 95%
Color Yellow
Quantity 5g
Chemical Name or Material N-Acetyl-L-alanine methyl ester

storageNote1 : -30° to -10°C