Alfa Aesar™ N-Acetyl-L-methionine, 99%

Manufacturer: Alfa Aesar™ B2431222

Quantity 100g

 View more versions of this product

Catalog No. AAB2431222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 65-82-7
    Molecular Formula C7H13NO3S
    Formula Weight 191.25
    MDL Number MFCD00064441
    Beilstein 1725552
    Merck Index 14,96
    Synonym Ac-Met-OH
    Chemical Name or Material N-Acetyl-L-methionine

    Specifications

    Formula Weight 191.25
    TSCA TSCA
    Molecular Formula C7H13NO3S
    MDL Number MFCD00064441
    CAS 65-82-7
    Beilstein 1725552
    Merck Index 14,96
    Synonym Ac-Met-OH
    Melting Point 103° to 107°C
    Quantity 100g
    Optical Rotation −21° (c=1.3 in Water)
    RTECS Number PD0480000
    Chemical Name or Material N-Acetyl-L-methionine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit