Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-tyrosine 99.0+%, TCI America™

Click to view available options
Quantity:
25g
537-55-3
C11H13NO4
223.228
MFCD00037190
CAHKINHBCWCHCF-JTQLQIEISA-N
n-acetyl-l-tyrosine,ac-tyr-oh,acetyl-l-tyrosine,n-acetyl-tyrosine,n-acetyltyrosine,acetyl tyrosine,l-n-acetyltyrosine,tyrosine, n-acetyl,l-tyrosine, n-acetyl,unii-da8g610zo5
68310
CHEBI:21563
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
537-55-3
C11H13NO4
223.228
MFCD00037190
CAHKINHBCWCHCF-JTQLQIEISA-N
n-acetyl-l-tyrosine,ac-tyr-oh,acetyl-l-tyrosine,n-acetyl-tyrosine,n-acetyltyrosine,acetyl tyrosine,l-n-acetyltyrosine,tyrosine, n-acetyl,l-tyrosine, n-acetyl,unii-da8g610zo5
68310
CHEBI:21563
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
Quantity 25g
Color White-Yellow
Melting Point 152°C
Formula Weight 223.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (T)
Chemical Name or Material N-Acetyl-L-tyrosine

EINECSNumber : (9)-1599

TSCA : Yes