Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-L-valine, 98%

Click to view available options
Quantity:
25g
100g
96-81-1
C7H13NO3
159.185
MFCD00066066
IHYJTAOFMMMOPX-LURJTMIESA-N
n-acetyl-l-valine,ac-val-oh,l-valine, n-acetyl,s-2-acetamido-3-methylbutanoic acid,acetyl-l-valine,n-acetylvaline,2s-2-acetamido-3-methylbutanoic acid,unii-u83p7h9hv3,acetylvaline,valine, n-acetyl-, l
66789
(2S)-2-acetamido-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)C
96-81-1
C7H13NO3
159.185
MFCD00066066
IHYJTAOFMMMOPX-LURJTMIESA-N
n-acetyl-l-valine,ac-val-oh,l-valine, n-acetyl,s-2-acetamido-3-methylbutanoic acid,acetyl-l-valine,n-acetylvaline,2s-2-acetamido-3-methylbutanoic acid,unii-u83p7h9hv3,acetylvaline,valine, n-acetyl-, l
66789
(2S)-2-acetamido-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)C
Beilstein 1723837
Formula Weight 159.19
Physical Form Crystalline Powder
Percent Purity 98%
Color White
Melting Point 163°C to 167°C
Quantity 100g
Chemical Name or Material N-Acetyl-L-valine

storageNote1 : -30° to -10°C