Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin

Click to view available options
Quantity:
1mg
5mg
292633-16-0
C33H41N7O11
711.729
MFCD01862664
ZILMZAQLFLKJPG-QORCZRPOSA-N
ac-leu-glu-his-asp-amc,ac-lehd-amc,ac-leu-glu-his-asp-7-amino-4-methylcoumarin,4s-4-1s-1-1s-2-carboxy-1-4-methyl-2-oxochromen-7-yl carbamoyl ethyl carbamoyl-2-1h-imidazol-4-yl ethyl carbamoyl-4-2s-2-acetamido-4-methylpentanamido butanoic acid
25108635
(4S)-4-[[(2S)-2-acetamido-4-methylpentanoyl]amino]-5-[[(2S)-1-[[(2S)-3-carboxy-1-[(4-methyl-2-oxochromen-7-yl)amino]-1-oxopropan-2-yl]amino]-3-(1H-imidazol-5-yl)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
CC1=CC(=O)OC2=C1C=CC(=C2)NC(=O)C(CC(=O)O)NC(=O)C(CC3=CN=CN3)NC(=O)C(CCC(=O)O)NC(=O)C(CC(C)C)NC(=O)C
292633-16-0
C33H41N7O11
711.729
MFCD01862664
ZILMZAQLFLKJPG-QORCZRPOSA-N
ac-leu-glu-his-asp-amc,ac-lehd-amc,ac-leu-glu-his-asp-7-amino-4-methylcoumarin,4s-4-1s-1-1s-2-carboxy-1-4-methyl-2-oxochromen-7-yl carbamoyl ethyl carbamoyl-2-1h-imidazol-4-yl ethyl carbamoyl-4-2s-2-acetamido-4-methylpentanamido butanoic acid
25108635
(4S)-4-[[(2S)-2-acetamido-4-methylpentanoyl]amino]-5-[[(2S)-1-[[(2S)-3-carboxy-1-[(4-methyl-2-oxochromen-7-yl)amino]-1-oxopropan-2-yl]amino]-3-(1H-imidazol-5-yl)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
CC1=CC(=O)OC2=C1C=CC(=C2)NC(=O)C(CC(=O)O)NC(=O)C(CC3=CN=CN3)NC(=O)C(CCC(=O)O)NC(=O)C(CC(C)C)NC(=O)C
Quantity 5mg
Color White
Formula Weight 711.72
Physical Form Lyophilized Powder
Chemical Name or Material N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin

storageNote1 : -30° to -10°C