Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-Leu-Glu-His-Asp-al

Click to view available options
Quantity:
1mg
5mg
MFCD01862665
Acetyl-LEHD-CHO; Caspase-9 Inhibitor
MFCD01862665
Acetyl-LEHD-CHO; Caspase-9 Inhibitor
Quantity 5mg
Color White
Sensitivity Light Sensitive
Formula Weight 538.55
Physical Form Lyophilized Powder
Chemical Name or Material N-Acetyl-Leu-Glu-His-Asp-al

storageNote1 : -30° to -10°C