Learn More
Tocris Bioscience™ N-Acetyl-N-acetoxy-4-chlorobenzenesulfonamide
Nitroxyl precursor
Manufacturer:  Tocris Bioscience™ 0722/10
 View more versions of this product
Catalog No. 07-221-0
Add to cart
Description
Description
A pro-drug of the potent vasorelaxant nitroxyl, H-N=O, which it slowly releases in neutral solution. HNO eventually dimerizes and dehydrates to N2O, without producing NO. Also a powerful inhibitor of aldehyde dehydrogenase.
Specifications
Specifications
142867-52-5
Observed MW: 291.71
Soluble to 100mM in DMSO
C10H10ClNO5S
10mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at +4°C