Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ N-Acetyl-N-acetoxy-4-chlorobenzenesulfonamide

$625.00

Specifications

Formula Weight Observed MW: 291.71
Molecular Formula C10H10ClNO5S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
07-225-0 View Documents Tocris Bioscience™
0722/50
50mg Each for $625.00
Add to cart
 
07-221-0 View Documents Tocris Bioscience™
0722/10
10mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

A pro-drug of the potent vasorelaxant nitroxyl, H-N=O, which it slowly releases in neutral solution. HNO eventually dimerizes and dehydrates to N2O, without producing NO. Also a powerful inhibitor of aldehyde dehydrogenase.
Specifications

Specifications

Observed MW: 291.71
C10H10ClNO5S
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at +4°C

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit