Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-O-tert-butyl-L-serine, 95%

Click to view available options
Quantity:
250mg
1g
5g
77285-09-7
C9H17NO4
203.238
MFCD00236770
COMGVZVKADFEPL-ZETCQYMHSA-N
ac-ser tbu-oh,l-serine,n-acetyl-o-1,1-dimethylethyl,ambotzaaa1904,n-acetyl-o-t-butyl-l-serine,2s-3-tert-butoxy-2-acetamidopropanoic acid
7019612
(2S)-2-acetamido-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(=O)NC(COC(C)(C)C)C(=O)O
77285-09-7
C9H17NO4
203.238
MFCD00236770
COMGVZVKADFEPL-ZETCQYMHSA-N
ac-ser tbu-oh,l-serine,n-acetyl-o-1,1-dimethylethyl,ambotzaaa1904,n-acetyl-o-t-butyl-l-serine,2s-3-tert-butoxy-2-acetamidopropanoic acid
7019612
(2S)-2-acetamido-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(=O)NC(COC(C)(C)C)C(=O)O
Beilstein 9047283
Formula Weight 203.24
Physical Form Crystalline Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material N-Acetyl-O-tert-butyl-L-serine

storageNote1 : -30° to -10°C