Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetyl-S-trityl-L-cysteine, 95%

Click to view available options
Quantity:
250mg
1g
5g
27486-87-9
C24H23NO3S
405.512
MFCD00236747
KCVPASSMLHHOIF-QFIPXVFZSA-N
ac-cys trt-oh,n-acetyl-s-trityl-l-cysteine,r-n-acetyl-s-tritylcysteine,2r-2-acetamido-3-triphenylmethyl sulfanyl propanoic acid,ac-cys trt-oh;,n-acetyl-s-trityl-l-cysteine;,l-cysteine,n-acetyl-s-triphenylmethyl,l-cysteine, n-acetyl-s-triphenylmethyl,s-trityl-l-cysteine stlc analogue, 37,2r-2-acetamido-3-tritylsulfanylpropanoic acid
7408363
(2R)-2-acetamido-3-tritylsulfanylpropanoic acid
CC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
27486-87-9
C24H23NO3S
405.512
MFCD00236747
KCVPASSMLHHOIF-QFIPXVFZSA-N
ac-cys trt-oh,n-acetyl-s-trityl-l-cysteine,r-n-acetyl-s-tritylcysteine,2r-2-acetamido-3-triphenylmethyl sulfanyl propanoic acid,ac-cys trt-oh;,n-acetyl-s-trityl-l-cysteine;,l-cysteine,n-acetyl-s-triphenylmethyl,l-cysteine, n-acetyl-s-triphenylmethyl,s-trityl-l-cysteine stlc analogue, 37,2r-2-acetamido-3-tritylsulfanylpropanoic acid
7408363
(2R)-2-acetamido-3-tritylsulfanylpropanoic acid
CC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
Beilstein 6877409
Formula Weight 405.51
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Acetyl-S-trityl-L-cysteine

storageNote1 : -30° to -10°C