Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-trans-4-hydroxy-L-proline, 99%, Alfa Aesar™

Click to view available options
Quantity:
10g
50g
33996-33-7
C7H11NO4
173.168
MFCD00037339
BAPRUDZDYCKSOQ-RITPCOANSA-N
oxaceprol,n-acetyl-l-hydroxyproline,2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid,oxaceprol inn,ac-hyp-oh,l-proline, 1-acetyl-4-hydroxy-, 4r,unii-q0xv76b96l,ac-hydroxyproline,trans-1-acetyl-4-hydroxy-l-proline,jonctum
65784
(2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
CC(=O)N1CC(CC1C(=O)O)O
33996-33-7
C7H11NO4
173.168
MFCD00037339
BAPRUDZDYCKSOQ-RITPCOANSA-N
oxaceprol,n-acetyl-l-hydroxyproline,2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid,oxaceprol inn,ac-hyp-oh,l-proline, 1-acetyl-4-hydroxy-, 4r,unii-q0xv76b96l,ac-hydroxyproline,trans-1-acetyl-4-hydroxy-l-proline,jonctum
65784
(2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
CC(=O)N1CC(CC1C(=O)O)O
Packaging Glass bottle
Formula Weight 173.17
Physical Form Crystalline Powder
Percent Purity 99%
Melting Point 131°C to 133°C
Quantity 50g
Chemical Name or Material N-Acetyl-trans-4-hydroxy-L-proline