Learn More
Alfa Aesar™ N-Acetyl-Tyr-Glu-Val-Asp-p-nitroanilide
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66566MB
 View more versions of this product
Catalog No. AAJ66566MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
686.66
25mg
Ac-YEVD-pNA
-30° to -10°C
N-Acetyl-Tyr-Glu-Val-Asp-p-nitroanilide
HPLC
Lyophilized powder
RUO
Caspase-1 Substrate I (ICE), chromogenic