Learn More
Alfa Aesar™ N-Acetyl-Val-Ala-Asp-al
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66696LB0
 View more versions of this product
Catalog No. AAJ66696LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
329.35
5mg
Acetyl-VAD-CHO
-30° to -10°C
HPLC
Lyophilized powder
RUO
N-Acetyl-Val-Ala-Asp-al