Learn More
Alfa Aesar™ N-Acetyl-Val-Asp-Val-Ala-Asp-7-amino-4-methylcoumarin
Manufacturer:  Alfa Aesar™ J66089MB
 View more versions of this product
Catalog No. AAJ66089MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
25mg
−30° to −10°C
C33H44N6O12
Ac-VDVAD-AMC; Caspase-2 Substrate (ICH-1) fluorogenic
White to Off-white
Lyophilized Powder
716.73g/mol
Signal Transduction Reagent