Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Acetylglucosaminyltransferase V/MGAT5 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | N-Acetylglucosaminyltransferase V/MGAT5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB022933
|
Novus Biologicals
NBP294188100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
N-Acetylglucosaminyltransferase V/MGAT5 Polyclonal antibody specifically detects N-Acetylglucosaminyltransferase V/MGAT5 in Human, Mouse samples. It is validated for Western BlotSpecifications
N-Acetylglucosaminyltransferase V/MGAT5 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
4249 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A, Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase, EC 2.4.1.155, GGNT5, GlcNAc-T V, GNT-VGNT-VA, Mannoside acetylglucosaminyltransferase 5, mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, N-acetylglucosaminyl-transferase V | |
Recombinant fusion protein containing a sequence corresponding to amino acids 627-741 of human MGAT5 (NP_002401.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title