Alfa Aesar™ N-Acetylglycinamide, 97%

Manufacturer: Alfa Aesar™ B2494003

Quantity 1g

 View more versions of this product

Catalog No. AAB2494003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2620-63-5
    Molecular Formula CH3CONHCH2CONH2
    Formula Weight 116.12
    MDL Number MFCD00008029
    Beilstein 1753982
    Synonym 2-Acetamidoacetamide; Ac-Gly-NH_2
    Chemical Name or Material N-Acetylglycinamide

    Specifications

    Formula Weight 116.12
    MDL Number MFCD00008029
    Beilstein 1753982
    RTECS Number MB7507000
    Odor Odorless
    Melting Point 136° to 140°C
    Quantity 1g
    Molecular Formula CH3CONHCH2CONH2
    CAS 2620-63-5
    Synonym 2-Acetamidoacetamide; Ac-Gly-NH_2
    Chemical Name or Material N-Acetylglycinamide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit