Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetylglycinamide, 97%

Click to view available options
Quantity:
1g
5g
25g
2620-63-5
C4H8N2O2
116.12
MFCD00008029
WQELDIQOHGAHEM-UHFFFAOYSA-N
n-acetylglycine amide,n-alpha-acetylglycinamide,unii-97sv9afw2z,acetamide, n-2-amino-2-oxoethyl,nalpha-acetylglycinamide,n-.alpha.-acetylglycinamide,97sv9afw2z,ethanimidic acid, n-2-amino-2-oxoethyl,acetylglutamide,acetylglycinamide
28326
2-acetamidoacetamide
CC(=O)NCC(=O)N
2620-63-5
C4H8N2O2
116.12
MFCD00008029
WQELDIQOHGAHEM-UHFFFAOYSA-N
n-acetylglycine amide,n-alpha-acetylglycinamide,unii-97sv9afw2z,acetamide, n-2-amino-2-oxoethyl,nalpha-acetylglycinamide,n-.alpha.-acetylglycinamide,97sv9afw2z,ethanimidic acid, n-2-amino-2-oxoethyl,acetylglutamide,acetylglycinamide
28326
2-acetamidoacetamide
CC(=O)NCC(=O)N
Quantity 25g
Melting Point 136°C to 140°C
Odor Odorless
Beilstein 1753982
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material N-Acetylglycinamide

RTECSNumber : MB7507000