Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Acetylglycyl-D-glutamic acid, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Excitatory peptide, more potent than L-glutamic acid at inducing seizures in mice.
Specifications
Formula Weight | 246.22 |
Percent Purity | >98% |
Quantity | 50 mg |
Chemical Name or Material | N-Acetylglycyl-D-glutamic acid |
Safety and Handling
Recommended Storage : Store at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title