Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Acetylglycylglycine, 97%

Click to view available options
Quantity:
250mg
1g
5687-48-9
C6H10N2O4
174.156
MFCD00037788
ZCASHLUDUSAKNN-UHFFFAOYSA-N
2-2-acetamidoacetamido acetic acid,n-acetylglycylglycine,ac-gly-gly-oh,unii-01u1cr2e6l,2-acetamidoacetamido acetic acid,n-ac-gly-gly,dpgt,acetyl-gly-gly,acetyl-glycylglycine,glycine, n-n-acetylglycyl
97591
2-[(2-acetamidoacetyl)amino]acetic acid
CC(=O)NCC(=O)NCC(=O)O
5687-48-9
C6H10N2O4
174.156
MFCD00037788
ZCASHLUDUSAKNN-UHFFFAOYSA-N
2-2-acetamidoacetamido acetic acid,n-acetylglycylglycine,ac-gly-gly-oh,unii-01u1cr2e6l,2-acetamidoacetamido acetic acid,n-ac-gly-gly,dpgt,acetyl-gly-gly,acetyl-glycylglycine,glycine, n-n-acetylglycyl
97591
2-[(2-acetamidoacetyl)amino]acetic acid
CC(=O)NCC(=O)NCC(=O)O
Beilstein 1782002
Formula Weight 174.16
Physical Form Solid
Percent Purity 97%
Quantity 1g
Chemical Name or Material N-Acetylglycylglycine