Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ N-Acetylmorpholine, 98%
Click to view available options
Quantity:
25g
100g
Specifications
Density | 1.116 |
Refractive Index | 1.4845 |
Boiling Point | 245°C to 246°C |
Quantity | 25g |
Flash Point | >110°C (230°F) |
Melting Point | 9°C to 10°C |
Beilstein | 111971 |
Formula Weight | 129.16 |
Percent Purity | 98% |
Chemical Name or Material | N-Acetylmorpholine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H360
EINECSNumber : 216-913-4
RTECSNumber : QD7000000
TSCA : Yes
Recommended Storage : Ambient temperatures