Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylneuraminic Acid, TCI America™

Click to view available options
Quantity:
100mg
1g
5g
131-48-6
C11H19NO9
309.271
MFCD00006620
SQVRNKJHWKZAKO-LFIUDZTESA-N
n-acetylneuramic acid,spectrum_000450,spectrum2_000559,spectrum4_000432,spectrum5_001326,spectrum2300147,2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
126963458
(4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
131-48-6
C11H19NO9
309.271
MFCD00006620
SQVRNKJHWKZAKO-LFIUDZTESA-N
n-acetylneuramic acid,spectrum_000450,spectrum2_000559,spectrum4_000432,spectrum5_001326,spectrum2300147,2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
126963458
(4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
Color White
Melting Point 186°C
Quantity 100mg
Formula Weight 309.27
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material N-Acetylneuraminic Acid

EINECSNumber : (5)-6979

TSCA : Yes

Recommended Storage : Freezer