Alfa Aesar™ N-Acetylthiourea, 98%

Manufacturer: Alfa Aesar™ 4700814

Quantity 25g

 View more versions of this product

Catalog No. AA4700814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 591-08-2
    Molecular Formula C3H6N2OS
    Formula Weight 118.16
    MDL Number MFCD00004937
    UN Number UN2811
    Beilstein 969960
    Chemical Name or Material N-Acetylthiourea

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 118.16
    MDL Number MFCD00004937
    CAS 591-08-2
    Beilstein 969960
    RTECS Number YR7700000
    Molecular Formula C3H6N2OS
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300
    Fatal if swallowed.
    Health Hazard 3 GHS P Statement
    P280f-P309-P310
    Wear protective clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Odor Odorless
    Melting Point 165° to 169°C
    Quantity 25g
    TSCA TSCA
    Color Pale cream
    Chemical Name or Material N-Acetylthiourea
    Assay 98%

    GHS H Statement
    H300
    Fatal if swallowed.

    GHS P Statement
    P280f-P309-P310
    Wear protective clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit